Assay Details
Target Gene Details
Entrez Gene ID: | 64426 |
Gene Name: | SDS3 homolog, SIN3A corepressor complex component |
Gene Aliases: |
SAP45, SDS3 |
Location: |
Chr.12:118373189-118418035 on Build GRCh38 |
Assay Gene Location: | Overlaps Exon 8 - Intron 8 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| SUDS3 | NM_022491.2 | NP_071936.2 | ||
| XM_011538671.2 | XP_011536973.1 | |||
| XM_017019817.1 | XP_016875306.1 | |||
| XM_017019818.1 | XP_016875307.1 | |||
| XM_017019819.1 | XP_016875308.1 | |||
| XM_017019820.1 | XP_016875309.1 | |||
| AK024460.1 | ||||
| BC098404.1 | AAH98404.1 | |||
| BX647687.1 |
More Information
Set Membership: |
|

Genomic Map