Assay Details
Target Gene Details
Entrez Gene ID: | 55922 |
Gene Name: | NFKB repressing factor |
Gene Aliases: |
ITBA4, NRF |
Location: |
Chr.X:119588337-119606464 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 1 - Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| NKRF | NM_001173487.1 | NP_001166958.1 | ||
| NM_017544.3 | NP_060014.2 | |||
| XM_011531365.2 | XP_011529667.1 | |||
| AL539002.3 | ||||
| AY208891.1 | AAP43025.1 | |||
| BC040379.1 | AAH40379.1 | |||
| BC047878.2 | AAH47878.1 | |||
| BC068514.1 | AAH68514.1 |
More Information
Set Membership: |
|

Genomic Map