Assay Details
Target Gene Details
Entrez Gene ID: | 54859 |
Gene Name: | elongator acetyltransferase complex subunit 6 |
Gene Aliases: |
C3orf75, TMEM103 |
Location: |
Chr.3:47495640-47513711 on Build GRCh38 |
Assay Gene Location: | Within Exon 11 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ELP6 | NM_001031703.2 | 7 | 1025 | NP_001026873.2 |
| XM_005265241.3 | 6 | 951 | XP_005265298.1 | |
| XM_017006669.1 | 7 | 1051 | XP_016862158.1 | |
| XM_017006670.1 | 7 | 1296 | XP_016862159.1 | |
| AK225721.1 | 7 | 1007 | ||
| AK298202.1 | 6 | 960 | ||
| BC000623.2 | 7 | 929 | AAH00623.1 | |
| BC014204.2 | 7 | 929 | AAH14204.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv3596083 | Chr.3:47485559 - 47529809 on Build GRCh38 | Gain |
|
| nsv428417 | Chr.3:46638962 - 47778557 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map