Assay Details
Target Gene Details
Entrez Gene ID: | 347734 |
Gene Name: | solute carrier family 35 member B2 |
Gene Aliases: |
PAPST1, SLL, UGTrel4 |
Location: |
Chr.6:44254101-44257890 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 3 - Exon 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| SLC35B2 | NM_001286509.1 | NP_001273438.1 | ||
| NM_001286510.1 | NP_001273439.1 | |||
| NM_001286511.1 | NP_001273440.1 | |||
| NM_001286512.1 | NP_001273441.1 | |||
| NM_001286513.1 | NP_001273442.1 | |||
| NM_001286517.1 | NP_001273446.1 | |||
| NM_001286519.1 | NP_001273448.1 | |||
| NM_001286520.1 | NP_001273449.1 | |||
| NM_178148.3 | NP_835361.1 | |||
| AB097021.1 | BAC77374.1 | |||
| AB097039.1 | BAC77392.1 | |||
| AB106538.1 | BAC79117.1 | |||
| AK075456.1 | BAC11631.1 | |||
| AK123749.1 | ||||
| AK293251.1 | ||||
| AK293344.1 | ||||
| AK307572.1 | ||||
| AY491520.1 | AAS79661.1 | |||
| BC024288.1 | AAH24288.1 | |||
| BI769501.1 | ||||
| CN370928.1 | ||||
| DA370522.1 | ||||
| DA383304.1 | ||||
| DA424392.1 | ||||
| DA817588.1 | ||||
| DB010939.1 | ||||
| HY044970.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv524398 | Chr.6:44233216 - 44286028 on Build GRCh38 | Loss |
|
| nsv5282 | Chr.6:44217138 - 44259154 on Build GRCh38 | Insertion |
|
| nsv603003 | Chr.6:44224183 - 44286028 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map