Assay Details
Target Gene Details
Entrez Gene ID: | 55016 |
Gene Name: | membrane associated ring-CH-type finger 1 |
Gene Aliases: |
MARCH-I, RNF171 |
Location: |
Chr.4:163524298-164384050 on Build GRCh38 |
Assay Gene Location: | Within Exon 12 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| MARCH1 | NM_001166373.1 | 8 | 1385 | NP_001159845.1 |
| NM_017923.3 | 4 | 1272 | NP_060393.1 | |
| XM_011532055.2 | 8 | 2090 | XP_011530357.1 | |
| XM_017008334.1 | 7 | 1312 | XP_016863823.1 | |
| XM_017008335.1 | 8 | 1691 | XP_016863824.1 | |
| XM_017008336.1 | 8 | 1557 | XP_016863825.1 | |
| AK000675.1 | 4 | 1161 | BAA91319.1 | |
| AK289711.1 | ||||
| AL713759.1 | 8 | 2180 | CAD28529.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1030184 | Chr.4:162402427 - 164455529 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map