Assay Details
Target Gene Details
Entrez Gene ID: | 56165 |
Gene Name: | tudor domain containing 1 |
Gene Aliases: |
CT41.1 |
Location: |
Chr.10:114174442-114232669 on Build GRCh38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| TDRD1 | XM_011539962.1 | 1 | 113 | XP_011538264.1 |
More Information
Set Membership: |
|

Genomic Map