Assay Details
Target Gene Details
Entrez Gene ID: | 9547 |
Gene Name: | C-X-C motif chemokine ligand 14 |
Gene Aliases: |
BMAC, BRAK, KEC, KS1, MIP-2g, MIP2G, NJAC, SCYB14 |
Location: |
Chr.5:135570679-135579279 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 3 - Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| CXCL14 | NM_004887.4 | NP_004878.2 | ||
| AF073957.1 | AAD03839.1 | |||
| AF086340.1 | ||||
| AF106911.1 | AAF78449.1 | |||
| AF144103.1 | AAD38944.1 | |||
| AK075514.1 | ||||
| AY358906.1 | AAQ89265.1 | |||
| BC003513.1 | AAH03513.1 | |||
| BT007080.1 | AAP35743.1 |
More Information
Set Membership: |
|

Genomic Map