Assay Details
Target Gene Details
Entrez Gene ID: | 64849 |
Gene Name: | solute carrier family 13 member 3 |
Gene Aliases: |
NADC3, SDCT2 |
Location: |
Chr.20:46557823-46684485 on Build GRCh38 |
Assay Gene Location: | Within Exon 15 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| SLC13A3 | NM_001011554.2 | 14 | 3234 | NP_001011554.1 |
| NM_001193339.1 | 12 | 3048 | NP_001180268.1 | |
| NM_001193340.1 | 13 | 3129 | NP_001180269.1 | |
| NM_001193342.1 | 13 | 3025 | NP_001180271.1 | |
| NM_022829.5 | 13 | 3198 | NP_073740.2 | |
| AF154121.1 | 13 | 3177 | AAF73251.1 | |
| AK056713.1 | 13 | 3196 | BAB71262.1 | |
| AK096658.1 | 13 | 3129 | BAC04834.1 | |
| AK123351.1 | 2 | 1822 | ||
| AL442082.1 | 4 | 1862 | CAC09447.2 | |
| AY072810.3 | 13 | 3177 | AAL66762.1 | |
| BC035966.1 | 13 | 3196 | AAH35966.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv833994 | Chr.20:46442116 - 46680210 on Build GRCh38 | Gain |
|
| nsv1061899 | Chr.20:46498425 - 47439490 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map