Assay Details
Target Gene Details
Entrez Gene ID: | 26050 |
Gene Name: | SLIT and NTRK like family member 5 |
Gene Aliases: |
LRRC11, bA364G4.2 |
Location: |
Chr.13:87671328-87679619 on Build GRCh38 |
Assay Gene Location: | Within Exon 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| SLITRK5 | NM_015567.1 | 2 | 4257 | NP_056382.1 |
| XM_005254038.4 | 2 | 4928 | XP_005254095.1 | |
| XM_005254039.2 | 2 | 4597 | XP_005254096.1 | |
| AB020725.1 | 2 | 4063 | ||
| AK024251.1 | 2 | 4257 | ||
| AW449813.1 | 1 | 205 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1044071 | Chr.13:86808989 - 88365666 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map