Assay Details
Target Gene Details
Entrez Gene ID: | 221044 |
Gene Name: | upper zone of growth plate and cartilage matrix associated |
Gene Aliases: |
C10orf49, GRP, GRP/UCMA |
Location: |
Chr.10:13221766-13234328 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 4 - Exon 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| UCMA | NM_001303118.1 | NP_001290047.1 | ||
| NM_001303119.1 | NP_001290048.1 | |||
| NM_145314.2 | NP_660357.2 | |||
| AA628504.1 | ||||
| BC018068.1 | AAH18068.2 | |||
| JX169863.1 | ||||
| JX169864.1 | ||||
| JX169865.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv971902 | Chr.10:13229614 - 13234697 on Build GRCh38 | Deletion |
|
| nsv528026 | Chr.10:13227931 - 13233643 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map