Assay Details
Target Gene Details
Entrez Gene ID: | 4794 |
Gene Name: | NFKB inhibitor epsilon |
Gene Aliases: |
IKBE |
Location: |
Chr.6:44258166-44265788 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 3 - Exon 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| NFKBIE | NM_004556.2 | NP_004547.2 | ||
| AK130739.1 | 1 | 329 | BAC85418.1 | |
| AK301360.1 | ||||
| BC011676.2 | AAH11676.2 | |||
| BC063609.1 | AAH63609.1 | |||
| U91616.1 | AAC51216.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv524398 | Chr.6:44233216 - 44286028 on Build GRCh38 | Loss |
|
| nsv603003 | Chr.6:44224183 - 44286028 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map