Assay Details
Target Gene Details
Entrez Gene ID: | 2281 |
Gene Name: | FK506 binding protein 1B |
Gene Aliases: |
FKBP12.6, FKBP1L, OTK4, PKBP1L, PPIase |
Location: |
Chr.2:24033205-24067743 on Build GRCh38 |
Assay Gene Location: | Overlaps - Exon 9 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| FKBP1B | XM_011532699.2 | XP_011531001.1 |
Target Gene Details
Entrez Gene ID: | 51639 |
Gene Name: | splicing factor 3b subunit 6 |
Gene Aliases: |
CGI-110, HSPC175, Ht006, P14, SAP14, SAP14a, SF3B14, SF3B14a |
Location: |
Chr.2:24067584-24076444 on Build GRCh38 |
Assay Gene Location: | Within Exon 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| SF3B6 | NM_016047.3 | 4 | 580 | NP_057131.1 |
| AF151868.1 | 4 | 576 | AAD34105.1 | |
| AF161523.1 | 4 | 474 | AAF29138.1 | |
| AF184213.1 | 4 | 576 | AAG09698.1 | |
| AF401310.1 | AAK94041.1 | |||
| AK000621.1 | 4 | 480 | ||
| AK311118.1 | 3 | 367 | ||
| AK311908.1 | ||||
| BC015463.1 | 4 | 580 | AAH15463.1 | |
| BG200650.1 | 3 | 341 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv997445 | Chr.2:23357900 - 24374347 on Build GRCh38 | Gain |
|
| nsv997725 | Chr.2:24052436 - 25123406 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map