Assay Details
Target Gene Details
Entrez Gene ID: | 79057 |
Gene Name: | proline rich and Gla domain 3 |
Gene Aliases: |
PRGP3, TMG3 |
Location: |
Chr.X:151694606-151701591 on Build GRCh38 |
Assay Gene Location: | Within Exon 6 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| PRRG3 | NM_024082.3 | 3 | 109 | NP_076987.3 |
| NR_033262.1 | 3 | 450 | ||
| XM_005274747.3 | 3 | 180 | XP_005274804.1 | |
| XM_011531197.2 | 3 | 229 | XP_011529499.1 | |
| AF326350.1 | 2 | 93 | AAK00955.1 | |
| AK074574.1 | 3 | 450 | BAC11069.1 | |
| BC128256.1 | 3 | 394 | ||
| BC128409.1 | 3 | 394 | ||
| CD631899.1 | 3 | 109 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv7156 | Chr.X:151673748 - 151705977 on Build GRCh38 | Insertion |
|
More Information
Set Membership: |
|

Genomic Map