Assay Details
Target Gene Details
Entrez Gene ID: | 54798 |
Gene Name: | dachsous cadherin-related 2 |
Gene Aliases: |
CDH27, CDHJ, CDHR7, PCDH23, PCDHJ |
Location: |
Chr.4:154234375-154491778 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 30 - Exon 31 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| DCHS2 | NM_017639.3 | NP_060109.2 | ||
| XM_011532045.1 | XP_011530347.1 | |||
| XM_011532046.2 | XP_011530348.1 | |||
| XM_011532049.2 | XP_011530351.1 | |||
| AF169691.1 | 1 | 514 | AAF89688.1 | |
| AK000054.1 | BAA90911.1 | |||
| AK095103.1 | ||||
| AK225200.1 | ||||
| AY354497.1 | AAR10443.1 | |||
| BX647835.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1023270 | Chr.4:154225348 - 154311669 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map