Assay Details
Target Gene Details
Entrez Gene ID: | 128372 |
Gene Name: | olfactory receptor family 6 subfamily N member 1 |
Gene Aliases: |
OR1-22 |
Location: |
Chr.1:158747733-158840979 on Build GRCh38 |
Assay Gene Location: | Within Intron 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| OR6N1 | XM_017000325.1 | XP_016855814.1 | ||
| XM_017000326.1 | XP_016855815.1 | |||
| XM_017000327.1 | XP_016855816.1 |
Target Gene Details
Entrez Gene ID: | 81442 |
Gene Name: | olfactory receptor family 6 subfamily N member 2 |
Gene Aliases: |
OR1-23 |
Location: |
Chr.1:158776682-158777635 on Build GRCh38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| OR6N2 | NM_001005278.1 | 1 | 449 | NP_001005278.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv997861 | Chr.1:158766425 - 158784871 on Build GRCh38 | Loss |
|
| dgv589n54 | Chr.1:158715246 - 158883163 on Build GRCh38 | Loss |
|
| nsv1002401 | Chr.1:158742837 - 159002241 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map