Assay Details
Target Gene Details
Entrez Gene ID: | 196951 |
Gene Name: | family with sequence similarity 227 member B |
Gene Aliases: |
C15orf33 |
Location: |
Chr.15:49326997-49620931 on Build GRCh38 |
Assay Gene Location: | Within Intron 23 |
Target Gene Details
Entrez Gene ID: | 2585 |
Gene Name: | galactokinase 2 |
Gene Aliases: |
GK2 |
Location: |
Chr.15:49155757-49367883 on Build GRCh38 |
Assay Gene Location: | Within Exon 14 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| GALK2 | NM_001001556.2 | 10 | 2926 | NP_001001556.1 |
| NM_001289030.1 | 11 | 3158 | NP_001275959.1 | |
| NM_001289031.1 | 10 | 3011 | NP_001275960.1 | |
| NM_002044.3 | 10 | 3028 | NP_002035.1 | |
| XM_005254279.3 | XP_005254336.1 | |||
| XM_005254280.3 | XP_005254337.1 | |||
| XM_005254284.3 | XP_005254341.1 | |||
| XM_006720461.3 | 11 | 2282 | XP_006720524.1 | |
| XM_006720462.3 | XP_006720525.1 | |||
| XM_006720463.3 | XP_006720526.1 | |||
| XM_006720464.3 | XP_006720527.1 | |||
| XM_011521441.2 | XP_011519743.1 | |||
| XM_017022062.1 | 9 | 2699 | XP_016877551.1 | |
| XM_017022063.1 | XP_016877552.1 | |||
| XM_017022064.1 | XP_016877553.1 | |||
| XM_017022065.1 | 12 | 2538 | XP_016877554.1 | |
| XM_017022066.1 | 12 | 2559 | XP_016877555.1 | |
| BX455251.2 | 1 | 298 | ||
| CA841825.1 | 1 | 303 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv833002 | Chr.15:49329203 - 49489602 on Build GRCh38 | Gain |
|
| esv3581695 | Chr.15:49319432 - 49331028 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map