Assay Details
Target Gene Details
Entrez Gene ID: | 116931 |
Gene Name: | mediator complex subunit 12 like |
Gene Aliases: |
NOPAR, TNRC11L, TRALP, TRALPUSH |
Location: |
Chr.3:151085665-151436677 on Build GRCh38 |
Assay Gene Location: | Within Intron 16 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| MED12L | NM_053002.5 | NP_443728.3 | ||
| XM_006713487.3 | XP_006713550.1 | |||
| XM_011512390.2 | XP_011510692.1 | |||
| XM_011512394.2 | XP_011510696.1 | |||
| XM_011512399.2 | XP_011510701.1 | |||
| XM_017005676.1 | XP_016861165.1 | |||
| XM_017005677.1 | XP_016861166.1 | |||
| XM_017005678.1 | XP_016861167.1 | |||
| XM_017005679.1 | XP_016861168.1 | |||
| XM_017005680.1 | XP_016861169.1 | |||
| AB046855.1 | BAB13461.1 | |||
| AF399708.1 | AAO38813.1 | |||
| AK128165.1 | ||||
| BC144559.1 | ||||
| BX108326.1 |
Target Gene Details
Entrez Gene ID: | 9934 |
Gene Name: | purinergic receptor P2Y14 |
Gene Aliases: |
BPR105, GPR105, P2Y14 |
Location: |
Chr.3:151212115-151279165 on Build GRCh38 |
Assay Gene Location: | Within Exon 6 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| P2RY14 | NM_001081455.1 | 3 | 1176 | NP_001074924.1 |
| NM_014879.3 | 3 | 986 | NP_055694.3 | |
| XM_005247922.3 | 4 | 965 | XP_005247979.1 | |
| XM_005247923.3 | 2 | 3169 | XP_005247980.1 | |
| XM_011513340.2 | 2 | 1601 | XP_011511642.1 | |
| XM_017007583.1 | 4 | 1771 | XP_016863072.1 | |
| AK289617.1 | 3 | 1080 | ||
| BC034989.2 | 3 | 910 | AAH34989.1 | |
| D13626.1 | 3 | 914 | ||
| EF577400.1 | 1 | 698 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv2759188 | Chr.3:151073838 - 151239697 on Build GRCh38 | Gain |
|
| nsv592045 | Chr.3:151052827 - 151377305 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map