Assay Details
Target Gene Details
Entrez Gene ID: | 29954 |
Gene Name: | protein O-mannosyltransferase 2 |
Gene Aliases: |
LGMD2N, MDDGA2, MDDGB2, MDDGC2 |
Location: |
Chr.14:77274956-77320885 on Build GRCh38 |
Assay Gene Location: | Within Exon 22 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| POMT2 | NM_013382.5 | 21 | 4365 | NP_037514.2 |
| XM_011536675.2 | 22 | 4554 | XP_011534977.1 | |
| XM_011536676.2 | 21 | 4467 | XP_011534978.1 | |
| XM_011536677.2 | 18 | 4096 | XP_011534979.1 | |
| XM_011536679.2 | 18 | 3756 | XP_011534981.1 | |
| XM_017021244.1 | 20 | 4199 | XP_016876733.1 | |
| XM_017021245.1 | 17 | 3653 | XP_016876734.1 | |
| AB209114.1 | 20 | 5140 | BAD92351.1 | |
| AI927974.1 | ||||
| AK023804.1 | 15 | 4956 | ||
| AL049462.1 | 1 | 156 | ||
| AL353956.1 | 4 | 2331 | CAB89256.1 | |
| AY090480.1 | 21 | 4368 | AAM12046.1 | |
| BM724049.1 | 1 | 584 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1361 | Chr.14:77241074 - 77286681 on Build GRCh38 | Deletion |
|
| nsv952872 | Chr.14:77274658 - 77279757 on Build GRCh38 | Deletion |
|
More Information
Set Membership: |
|

Genomic Map