Assay Details
Target Gene Details
Entrez Gene ID: | 2959 |
Gene Name: | general transcription factor IIB |
Gene Aliases: |
TF2B, TFIIB |
Location: |
Chr.1:88852638-88891944 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 4 - Exon 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| GTF2B | NM_001514.5 | NP_001505.1 | ||
| XM_011541299.1 | XP_011539601.1 | |||
| AB451296.1 | ||||
| AK289822.1 | ||||
| BC020597.1 | AAH20597.1 | |||
| BP379342.1 | 4 | 497 | ||
| M76766.1 | AAA61149.1 | |||
| S44184.1 | AAB23144.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv830537 | Chr.1:88790384 - 88940395 on Build GRCh38 | Gain |
|
| nsv830525 | Chr.1:88776863 - 88975647 on Build GRCh38 | Loss |
|
| nsv830514 | Chr.1:88715143 - 88876830 on Build GRCh38 | Loss |
|
| dgv22n27 | Chr.1:88791695 - 88894837 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map