Assay Details
Target Gene Details
Entrez Gene ID: | 4548 |
Gene Name: | 5-methyltetrahydrofolate-homocysteine methyltransferase |
Gene Aliases: |
HMAG, MS, cblG |
Location: |
Chr.1:236794304-236903981 on Build GRCh38 |
Assay Gene Location: | Within Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| MTR | NM_000254.2 | 2 | 515 | NP_000245.2 |
| NM_001291939.1 | 2 | 515 | NP_001278868.1 | |
| NM_001291940.1 | 2 | 515 | NP_001278869.1 | |
| XM_005273141.4 | 2 | 484 | XP_005273198.1 | |
| XM_011544194.2 | XP_011542496.1 | |||
| XM_017001329.1 | XP_016856818.1 | |||
| XM_017001330.1 | XP_016856819.1 | |||
| AK294270.1 | ||||
| AK299904.1 | 2 | 515 | ||
| AK307390.1 | 2 | 504 | ||
| BC130616.1 | 2 | 114 | ||
| BC136440.1 | 2 | 114 | ||
| BC144095.1 | 2 | 114 | ||
| BC144097.1 | 2 | 114 | ||
| DC377743.1 | 2 | 515 | ||
| U71285.1 | 2 | 155 | AAC51188.1 | |
| U73338.1 | 2 | 486 | AAB39704.1 | |
| U75743.1 | 2 | 378 | AAB58906.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1005877 | Chr.1:235912335 - 237038508 on Build GRCh38 | Gain |
|
| nsv5010 | Chr.1:236796351 - 236810727 on Build GRCh38 | Insertion |
|
| nsv428335 | Chr.1:236734659 - 236913672 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map