Assay Details
Target Gene Details
Entrez Gene ID: | 9063 |
Gene Name: | protein inhibitor of activated STAT 2 |
Gene Aliases: |
ARIP3, DIP, MIZ, MIZ1, PIASX, PIASX-ALPHA, PIASX-BETA, SIZ2, ZMIZ4 |
Location: |
Chr.18:46803224-46920167 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 18 - Exon 19 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv9629 | Chr.18:46684730 - 47231140 on Build GRCh38 | Gain+Loss |
|
| dgv614e214 | Chr.18:46702717 - 46833888 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map