Assay Details
Target Gene Details
Entrez Gene ID: | 23051 |
Gene Name: | zinc fingers and homeoboxes 3 |
Gene Aliases: |
TIX1 |
Location: |
Chr.20:41178449-41317606 on Build GRCh38 |
Assay Gene Location: | Within Exon 16 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ZHX3 | NM_015035.3 | 4 | 5946 | NP_055850.1 |
| XM_005260341.3 | 4 | 5747 | XP_005260398.1 | |
| XM_011528720.1 | 4 | 5981 | XP_011527022.1 | |
| XM_017027737.1 | 4 | 5989 | XP_016883226.1 | |
| XM_017027738.1 | 4 | 6112 | XP_016883227.1 | |
| AB007855.3 | 4 | 5917 | ||
| AB081948.1 | 4 | 5928 | BAC65211.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1064246 | Chr.20:40261343 - 42254320 on Build GRCh38 | Loss |
|
| nsv833983 | Chr.20:41051961 - 41213015 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map