Assay Details
Target Gene Details
Entrez Gene ID: | 2827 |
Gene Name: | G protein-coupled receptor 3 |
Gene Aliases: |
ACCA |
Location: |
Chr.1:27392644-27395814 on Build GRCh38 |
Assay Gene Location: | Within Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| GPR3 | NM_005281.3 | 2 | 274 | NP_005272.1 |
| AK291185.1 | 2 | 261 | ||
| AK314868.1 | 2 | 253 | ||
| BC032702.1 | 2 | 270 | AAH32702.1 | |
| BQ882813.1 | 2 | 274 | ||
| HQ709189.1 | 1 | 175 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv526051 | Chr.1:27284255 - 27445496 on Build GRCh38 | Gain |
|
| nsv950342 | Chr.1:27392797 - 27410906 on Build GRCh38 | Deletion |
|
| nsv516766 | Chr.1:27335744 - 27405147 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map