Assay Details
Target Gene Details
Entrez Gene ID: | 284525 |
Gene Name: | solute carrier family 9 member C2 (putative) |
Gene Aliases: |
SLC9A11 |
Location: |
Chr.1:173500465-173603094 on Build GRCh38 |
Assay Gene Location: | Within Exon 25 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| SLC9C2 | NM_178527.3 | 25 | 3507 | NP_848622.2 |
| XM_011509426.1 | 25 | 3507 | XP_011507728.1 | |
| XM_017001065.1 | 25 | 3202 | XP_016856554.1 | |
| XM_017001066.1 | 25 | 3196 | XP_016856555.1 | |
| XM_017001067.1 | 24 | 3387 | XP_016856556.1 | |
| XM_017001068.1 | 24 | 3406 | XP_016856557.1 | |
| XM_017001069.1 | 24 | 3101 | XP_016856558.1 | |
| XM_017001070.1 | 24 | 3095 | XP_016856559.1 | |
| XM_017001071.1 | 25 | 3114 | XP_016856560.1 | |
| XM_017001072.1 | 23 | 3240 | XP_016856561.1 | |
| XM_017001073.1 | 24 | 3013 | XP_016856562.1 | |
| XM_017001074.1 | 23 | 2942 | XP_016856563.1 | |
| XM_017001075.1 | 18 | 2269 | XP_016856564.1 | |
| XM_017001076.1 | 15 | 1876 | XP_016856565.1 | |
| AK128104.1 | 25 | 3507 | ||
| BC042592.1 | 25 | 3454 | AAH42592.1 | |
| BX648119.1 | 18 | 2418 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv831915 | Chr.1:173299285 - 173513018 on Build GRCh38 | Gain+Loss |
|
More Information
Set Membership: |
|

Genomic Map