Assay Details
Target Gene Details
Entrez Gene ID: | 83604 |
Gene Name: | transmembrane protein 47 |
Gene Aliases: |
BCMP1, TM4SF10 |
Location: |
Chr.X:34627064-34657288 on Build GRCh38 |
Assay Gene Location: | Within Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| TMEM47 | NM_031442.3 | 3 | 3729 | NP_113630.1 |
| AL049257.1 | 1 | 2151 | ||
| AL136550.1 | 3 | 3479 | CAB66485.1 | |
| BC039242.1 | 3 | 3729 | AAH39242.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv2664433 | Chr.X:34458727 - 34946283 on Build GRCh38 | Deletion |
|
| nsv519268 | Chr.X:34474253 - 34646918 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map