Assay Details
Target Gene Details
Entrez Gene ID: | 8936 |
Gene Name: | WAS protein family member 1 |
Gene Aliases: |
SCAR1, WAVE, WAVE1 |
Location: |
Chr.6:110099817-110189874 on Build GRCh38 |
Assay Gene Location: | Within Exon 11 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| WASF1 | NM_001024934.1 | 7 | 1366 | NP_001020105.1 |
| NM_001024935.1 | 7 | 1319 | NP_001020106.1 | |
| NM_001024936.1 | 6 | 1221 | NP_001020107.1 | |
| NM_003931.2 | 8 | 1464 | NP_003922.1 | |
| XM_005267203.3 | 9 | 1462 | XP_005267260.1 | |
| XM_005267205.2 | 7 | 928 | XP_005267262.1 | |
| XM_005267206.2 | 7 | 976 | XP_005267263.1 | |
| XM_005267207.2 | 6 | 791 | XP_005267264.1 | |
| XM_011536233.1 | 7 | 1076 | XP_011534535.1 | |
| XM_011536234.1 | 8 | 1364 | XP_011534536.1 | |
| XM_011536235.1 | 8 | 1419 | XP_011534537.1 | |
| XM_017011437.1 | 9 | 1484 | XP_016866926.1 | |
| XM_017011438.1 | 9 | 1426 | XP_016866927.1 | |
| XM_017011439.1 | 7 | 1014 | XP_016866928.1 | |
| XM_017011440.1 | 6 | 891 | XP_016866929.1 | |
| XM_017011441.1 | 7 | 925 | XP_016866930.1 | |
| XM_017011442.1 | 7 | 1219 | XP_016866931.1 | |
| XM_017011443.1 | 6 | 821 | XP_016866932.1 | |
| XM_017011444.1 | 7 | 973 | XP_016866933.1 | |
| AF134303.1 | 5 | 627 | AAD33052.1 | |
| AK097838.1 | ||||
| BC044591.1 | 7 | 937 | AAH44591.1 | |
| D87459.1 | 7 | 869 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv2759462 | Chr.6:110068723 - 110183477 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map