Assay Details
Target Gene Details
Entrez Gene ID: | 283635 |
Gene Name: | family with sequence similarity 177 member A1 |
Gene Aliases: |
C14orf24 |
Location: |
Chr.14:35044907-35083383 on Build GRCh38 |
Assay Gene Location: | Within Exon 7 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| FAM177A1 | NM_001079519.1 | 7 | 1569 | NP_001072987.1 |
| NM_001289022.1 | 6 | 1292 | NP_001275951.1 | |
| NM_173607.3 | 5 | 1239 | NP_775878.2 | |
| AK096173.1 | 6 | 1180 | BAC04716.1 | |
| AL833508.1 | 5 | 1153 | ||
| BX640884.1 | 5 | 1242 | ||
| BX640984.1 | 5 | 1190 | ||
| CR749796.1 | 5 | 1239 | CAH18656.1 |
Target Gene Details
Entrez Gene ID: | 101927178 |
Gene Name: | uncharacterized LOC101927178 |
Gene Aliases: |
- |
Location: |
Chr.14:35081081-35113130 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| LOC101927178 | NR_110415.1 | |||
| CK904250.1 | ||||
| CK904251.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv2515788 | Chr.14:35080978 - 35082597 on Build GRCh38 | Deletion |
|
| nsv564263 | Chr.14:34997378 - 35087292 on Build GRCh38 | Loss |
|
| esv3892499 | Chr.14:34900273 - 35107577 on Build GRCh38 | Loss |
|
| nsv564264 | Chr.14:35013591 - 35263163 on Build GRCh38 | Gain |
|
| nsv564265 | Chr.14:35043129 - 35155625 on Build GRCh38 | Loss |
|
| dgv1868n100 | Chr.14:35041579 - 35287111 on Build GRCh38 | Gain |
|
| esv3584249 | Chr.14:35060670 - 35255038 on Build GRCh38 | Gain |
|
| nsv1042625 | Chr.14:35076888 - 35211152 on Build GRCh38 | Gain |
|

Genomic Map