Assay Details
Target Gene Details
Entrez Gene ID: | 1393 |
Gene Name: | corticotropin releasing hormone binding protein |
Gene Aliases: |
CRF-BP, CRFBP |
Location: |
Chr.5:76952713-76981143 on Build GRCh38 |
Assay Gene Location: | Within Exon 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| CRHBP | NM_001882.3 | 5 | 922 | NP_001873.2 |
| AK223457.1 | 5 | 799 | BAD97177.1 | |
| AK307729.1 | 5 | 732 | ||
| AK311052.1 | 5 | 728 | ||
| BC018038.1 | 5 | 732 | AAH18038.1 | |
| CR541666.1 | 5 | 642 | CAG46467.1 | |
| X58022.1 | 5 | 688 | CAA41086.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv830355 | Chr.5:76898897 - 77072195 on Build GRCh38 | Gain |
|
| nsv1027236 | Chr.5:76895111 - 77118908 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map