Assay Details
Target Gene Details
Entrez Gene ID: | 3604 |
Gene Name: | TNF receptor superfamily member 9 |
Gene Aliases: |
4-1BB, CD137, CDw137, ILA |
Location: |
Chr.1:7915871-7943165 on Build GRCh38 |
Assay Gene Location: | Within Exon 10 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| TNFRSF9 | NM_001561.5 | 9 | 1682 | NP_001552.2 |
| XM_011541386.2 | 8 | 1697 | XP_011539688.1 | |
| BC006196.2 | 8 | 1542 | AAH06196.1 |
More Information
Set Membership: |
|

Genomic Map