Assay Details
Target Gene Details
Entrez Gene ID: | 23318 |
Gene Name: | zinc finger CCHC-type containing 11 |
Gene Aliases: |
PAPD3, TUT4 |
Location: |
Chr.1:52423275-52553463 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 26 - Exon 26 |
More Information
Set Membership: |
|

Genomic Map