Assay Details
Target Gene Details
Entrez Gene ID: | 83988 |
Gene Name: | neurocalcin delta |
Gene Aliases: |
- |
Location: |
Chr.8:101686542-102124907 on Build GRCh38 |
Assay Gene Location: | Within Exon 15 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| NCALD | NM_001040624.1 | 8 | 1738 | NP_001035714.1 |
| NM_001040625.1 | 7 | 1651 | NP_001035715.1 | |
| NM_001040626.1 | 7 | 1651 | NP_001035716.1 | |
| NM_001040627.1 | 6 | 1564 | NP_001035717.1 | |
| NM_001040628.1 | 8 | 1707 | NP_001035718.1 | |
| NM_001040629.1 | 7 | 1620 | NP_001035719.1 | |
| NM_001040630.1 | 6 | 1804 | NP_001035720.1 | |
| NM_032041.2 | 4 | 1713 | NP_114430.2 | |
| XM_011517332.2 | 4 | 1502 | XP_011515634.1 | |
| XM_011517333.2 | 8 | 1649 | XP_011515635.1 | |
| XM_011517334.2 | 5 | 1533 | XP_011515636.1 | |
| XM_011517335.2 | 6 | 1625 | XP_011515637.1 | |
| XM_017013901.1 | 7 | 1841 | XP_016869390.1 | |
| XM_017013902.1 | 5 | 1512 | XP_016869391.1 | |
| AB209015.1 | 5 | 4529 | BAD92252.1 | |
| AF251061.1 | 6 | 1492 | AAK34951.1 | |
| AK055073.1 | 4 | 1471 | ||
| AK055097.1 | 2 | 2441 | ||
| AK096515.1 | 8 | 1663 | ||
| AL136612.1 | 4 | 1464 | CAB66547.1 | |
| BC036098.1 | 6 | 1491 | AAH36098.1 | |
| BC063428.1 | AAH63428.1 |
More Information
Set Membership: |
|

Genomic Map