Assay Details
Target Gene Details
Entrez Gene ID: | 10432 |
Gene Name: | RNA binding motif protein 14 |
Gene Aliases: |
COAA, PSP2, SIP, SYTIP1, TMEM137 |
Location: |
Chr.11:66616582-66629934 on Build GRCh38 |
Assay Gene Location: | Within Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| RBM14 | NM_001198836.1 | NP_001185765.1 | ||
| NM_001198837.1 | NP_001185766.1 | |||
| NM_006328.3 | 2 | 918 | NP_006319.1 | |
| AB209007.1 | 1 | 3619 | BAD92244.1 | |
| AF080561.1 | 2 | 859 | AAC64058.1 | |
| AF315632.1 | 2 | 874 | AAK77961.1 | |
| AF315633.1 | AAK77962.1 | |||
| AK021768.1 | 2 | 870 | ||
| AK222830.1 | 2 | 871 | BAD96550.1 | |
| AK297907.1 | ||||
| AK307254.1 | ||||
| BC000488.2 | 2 | 854 | AAH00488.1 | |
| DQ294957.1 | ABB99396.1 | |||
| KU178520.1 | 2 | 779 | ||
| KU178521.1 | ||||
| KU178522.1 |
Target Gene Details
Entrez Gene ID: | 100526737 |
Gene Name: | RBM14-RBM4 readthrough |
Gene Aliases: |
COAZ |
Location: |
Chr.11:66616582-66646473 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| RBM14-RBM4 | NM_001198845.1 | NP_001185774.1 | ||
| NM_001198846.1 | NP_001185775.1 | |||
| AW958943.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv522277 | Chr.11:66466552 - 66726354 on Build GRCh38 | Loss |
|
| nsv363 | Chr.11:66597906 - 66629110 on Build GRCh38 | Insertion |
|
More Information
Set Membership: |
|

Genomic Map