Assay Details
Target Gene Details
Entrez Gene ID: | 23300 |
Gene Name: | ATM interactor |
Gene Aliases: |
ASCIZ, ZNF822 |
Location: |
Chr.16:81035853-81047346 on Build GRCh38 |
Assay Gene Location: | Within Exon 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ATMIN | NM_001300728.1 | 4 | 2766 | NP_001287657.1 |
| NM_015251.2 | 4 | 2826 | NP_056066.2 | |
| AB007891.1 | 3 | 3303 | ||
| AK025058.1 | 1 | 664 | ||
| CR749457.1 | 4 | 2719 | CAH18291.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv573338 | Chr.16:80986788 - 81073339 on Build GRCh38 | Loss |
|
| nsv1897 | Chr.16:81026961 - 81071655 on Build GRCh38 | Deletion |
|
| nsv1062724 | Chr.16:81018707 - 81103402 on Build GRCh38 | Gain |
|
| esv2422360 | Chr.16:81032428 - 81147122 on Build GRCh38 | Duplication |
|
More Information
Set Membership: |
|

Genomic Map