Assay Details
Target Gene Details
Entrez Gene ID: | 84106 |
Gene Name: | PML-RARA regulated adaptor molecule 1 |
Gene Aliases: |
PML-RAR, PRAM-1 |
Location: |
Chr.19:8490055-8502654 on Build GRCh38 |
Assay Gene Location: | Within Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| PRAM1 | NM_032152.4 | 2 | 642 | NP_115528.4 |
| XM_005272502.2 | 2 | 628 | XP_005272559.1 | |
| XM_011528352.2 | 2 | 648 | XP_011526654.1 | |
| XM_011528353.2 | 2 | 646 | XP_011526655.1 | |
| AJ272324.1 | 1 | 786 | CAC17767.1 | |
| AK308227.1 | 2 | 625 | ||
| BC028012.1 | 3 | 599 | AAH28012.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| dgv137n111 | Chr.19:8488438 - 8590905 on Build GRCh38 | Deletion |
|
| dgv6258n54 | Chr.19:8487389 - 8564616 on Build GRCh38 | Loss |
|
| nsv578559 | Chr.19:8483291 - 8532981 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map