Assay Details
Target Gene Details
Entrez Gene ID: | 56652 |
Gene Name: | chromosome 10 open reading frame 2 |
Gene Aliases: |
ATXN8, IOSCA, MTDPS7, PEO, PEO1, PEOA3, PRLTS5, SANDO, SCA8, TWINL |
Location: |
Chr.10:100987527-100994403 on Build GRCh38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| C10orf2 | NM_001163812.1 | 1 | 1293 | NP_001157284.1 |
| NM_001163813.1 | NP_001157285.1 | |||
| NM_001163814.1 | NP_001157286.1 | |||
| NM_021830.4 | 1 | 1293 | NP_068602.2 | |
| XM_011539975.2 | XP_011538277.1 | |||
| XM_017016437.1 | 1 | 792 | XP_016871926.1 | |
| AF292004.1 | 1 | 803 | AAK69558.1 | |
| AF292005.1 | 1 | 803 | AAK69559.1 | |
| AK025485.1 | 1 | 116 | BAB15148.1 | |
| AK297068.1 | ||||
| AK308098.1 | 1 | 1293 | ||
| BC013349.2 | 1 | 226 | AAH13349.1 | |
| BC033762.1 | 1 | 286 | AAH33762.1 | |
| BQ643363.1 | ||||
| BX640829.1 | 1 | 1275 | CAE45905.1 | |
| CB112401.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv831965 | Chr.10:100916724 - 101082796 on Build GRCh38 | Loss |
|
| nsv517698 | Chr.10:100965896 - 101017113 on Build GRCh38 | Loss |
|
| nsv467443 | Chr.10:100929460 - 101007360 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map