Assay Details
Target Gene Details
Entrez Gene ID: | 60481 |
Gene Name: | ELOVL fatty acid elongase 5 |
Gene Aliases: |
HELO1, SCA38, dJ483K16.1 |
Location: |
Chr.6:53267398-53349179 on Build GRCh38 |
Assay Gene Location: | Within Exon 10 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ELOVL5 | NM_001242828.1 | 9 | 2654 | NP_001229757.1 |
| NM_001242830.1 | 7 | 2448 | NP_001229759.1 | |
| NM_001301856.1 | 8 | 2806 | NP_001288785.1 | |
| NM_021814.4 | 8 | 2573 | NP_068586.1 | |
| AB209798.1 | 7 | 2196 | BAD93035.1 | |
| AF052129.1 | 1 | 762 | ||
| AF111849.1 | 2 | 1593 | AAF16688.1 | |
| AF338241.1 | 8 | 2546 | AAM00193.1 | |
| AK074748.1 | 3 | 1670 | BAC11178.1 | |
| AK074889.1 | 7 | 2207 | BAC11270.1 | |
| AK125098.1 | 8 | 2650 | ||
| AL136939.1 | 8 | 2321 | CAB66873.1 | |
| BC017270.2 | 3 | 1617 | AAH17270.2 | |
| BC067123.1 | 8 | 2322 | AAH67123.2 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1033994 | Chr.6:53225739 - 53458464 on Build GRCh38 | Gain |
|
| esv2763940 | Chr.6:53228517 - 53622284 on Build GRCh38 | Gain+Loss |
|
More Information
Set Membership: |
|

Genomic Map