Assay Details
Target Gene Details
Entrez Gene ID: | 22930 |
Gene Name: | RAB3 GTPase activating protein catalytic subunit 1 |
Gene Aliases: |
P130, RAB3GAP, RAB3GAP130, WARBM1 |
Location: |
Chr.2:135052265-135176667 on Build GRCh38 |
Assay Gene Location: | Within Exon 25 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| RAB3GAP1 | NM_001172435.1 | 25 | 3155 | NP_001165906.1 |
| NM_012233.2 | 24 | 3134 | NP_036365.1 | |
| XM_011510823.2 | XP_011509125.1 | |||
| XM_011510825.2 | XP_011509127.1 | |||
| AK022494.1 | 24 | 3106 | ||
| AK056058.1 | 2 | 1042 | ||
| AK309118.1 | 7 | 1036 | ||
| AL096752.1 | CAH56411.1 | |||
| BC022977.1 | 11 | 1778 | AAH22977.1 | |
| BC146809.1 | ||||
| D31886.1 | 24 | 3093 | BAA06684.1 |
Target Gene Details
Entrez Gene ID: | 84083 |
Gene Name: | zinc finger RANBP2-type containing 3 |
Gene Aliases: |
4933425L19Rik, AH2 |
Location: |
Chr.2:135164703-135531322 on Build GRCh38 |
Assay Gene Location: | Within Intron 25 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ZRANB3 | XM_011511959.2 | XP_011510261.2 | ||
| XM_011511961.2 | XP_011510263.2 | |||
| XM_011511966.2 | XP_011510268.2 | |||
| XM_017005065.1 | XP_016860554.1 | |||
| XM_017005066.1 | XP_016860555.1 | |||
| XM_017005067.1 | XP_016860556.1 | |||
| XM_017005068.1 | XP_016860557.1 | |||
| XM_017005069.1 | XP_016860558.1 | |||
| XM_017005070.1 | XP_016860559.1 | |||
| XM_017005071.1 | XP_016860560.1 | |||
| AL136824.1 | CAB66758.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv834386 | Chr.2:135110881 - 135223815 on Build GRCh38 | Gain |
|
| esv3592568 | Chr.2:134994189 - 135317860 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map