Assay Details
Target Gene Details
Entrez Gene ID: | 83989 |
Gene Name: | family with sequence similarity 172 member A |
Gene Aliases: |
C5orf21 |
Location: |
Chr.5:93615130-94111699 on Build GRCh38 |
Assay Gene Location: | Within Exon 21 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| FAM172A | NM_001163417.1 | 10 | 1527 | NP_001156889.1 |
| NM_001163418.1 | 9 | 1335 | NP_001156890.1 | |
| NM_032042.5 | 11 | 1651 | NP_114431.2 | |
| NR_028080.1 | 9 | 1358 | ||
| XM_005272105.3 | 10 | 1424 | XP_005272162.1 | |
| XM_005272106.3 | 10 | 1398 | XP_005272163.1 | |
| XM_005272108.3 | 10 | 1545 | XP_005272165.1 | |
| XM_005272110.3 | 9 | 1252 | XP_005272167.1 | |
| XM_006714717.3 | 11 | 1643 | XP_006714780.1 | |
| XM_006714718.3 | 9 | 2257 | XP_006714781.1 | |
| XM_006714719.3 | 9 | 1406 | XP_006714782.1 | |
| XM_011543672.2 | 11 | 1547 | XP_011541974.1 | |
| XM_011543673.2 | 10 | 1446 | XP_011541975.1 | |
| XM_017009952.1 | 11 | 1939 | XP_016865441.1 | |
| XM_017009953.1 | 11 | 1937 | XP_016865442.1 | |
| XM_017009955.1 | 10 | 2355 | XP_016865444.1 | |
| XM_017009956.1 | 10 | 1669 | XP_016865445.1 | |
| XM_017009957.1 | 9 | 1623 | XP_016865446.1 | |
| XM_017009958.1 | 9 | 1418 | XP_016865447.1 | |
| XM_017009960.1 | 8 | 1200 | XP_016865449.1 | |
| AK295879.1 | 9 | 1284 | ||
| AK296683.1 | 5 | 836 | ||
| AK309742.1 | 2 | 437 | ||
| BC020584.2 | 10 | 1453 | AAH20584.1 |
Target Gene Details
Entrez Gene ID: | 100313887 |
Gene Name: | microRNA 2277 |
Gene Aliases: |
mir-2277 |
Location: |
Chr.5:93620696-93620788 on Build GRCh38 |
Assay Gene Location: | Overlaps - Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| MIR2277 | NR_031754.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| dgv954n67 | Chr.5:93560640 - 93633962 on Build GRCh38 | Gain |
|
| nsv830404 | Chr.5:93532630 - 93712551 on Build GRCh38 | Loss |
|

Genomic Map