Assay Details
Target Gene Details
Entrez Gene ID: | 3082 |
Gene Name: | hepatocyte growth factor |
Gene Aliases: |
DFNB39, F-TCF, HGFB, HPTA, SF |
Location: |
Chr.7:81699006-81770438 on Build GRCh38 |
Assay Gene Location: | Within Exon 20 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| HGF | NM_000601.5 | 18 | 2373 | NP_000592.3 |
| NM_001010932.2 | 18 | 2358 | NP_001010932.1 | |
| XM_006715956.2 | 19 | 2336 | XP_006716019.1 | |
| XM_011516115.2 | 19 | 2321 | XP_011514417.1 | |
| AK291674.1 | 18 | 2308 | ||
| AK313694.1 | ||||
| BC130284.1 | 18 | 2221 | ||
| BC130286.1 | 18 | 2221 | ||
| M29145.1 | 18 | 2247 | AAA52650.1 | |
| M55379.1 | ||||
| M60718.1 | 18 | 2218 | AAA52648.1 | |
| M73239.1 | 18 | 2211 | AAA64239.1 | |
| M73240.1 | 18 | 2196 | AAA64297.1 | |
| X16323.1 | 18 | 2280 | CAA34387.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv3576398 | Chr.7:81685626 - 81732874 on Build GRCh38 | Gain |
|
| nsv1019753 | Chr.7:81692852 - 82052256 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map