Assay Details
Target Gene Details
Entrez Gene ID: | 64430 |
Gene Name: | pecanex homolog 4 (Drosophila) |
Gene Aliases: |
C14orf135, FBP2, PCNXL4 |
Location: |
Chr.14:60091276-60142976 on Build GRCh38 |
Assay Gene Location: | Within Exon 9 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| PCNX4 | NM_022495.5 | 8 | 1942 | NP_071940.4 |
| XM_005268009.2 | 9 | 3319 | XP_005268066.1 | |
| AK023008.1 | 3 | 430 | ||
| AK095489.1 | 8 | 1926 | ||
| BX648440.1 | 4 | 666 | CAH56141.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1044390 | Chr.14:60109151 - 60168783 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map