Assay Details
Target Gene Details
Entrez Gene ID: | 8653 |
Gene Name: | DEAD-box helicase 3, Y-linked |
Gene Aliases: |
DBY |
Location: |
Chr.Y:12903999-12920478 on Build GRCh38 |
Assay Gene Location: | Within Exon 7 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| DDX3Y | NM_001122665.3 | NP_001116137.1 | ||
| NM_001302552.2 | NP_001289481.1 | |||
| NM_001324195.1 | NP_001311124.1 | |||
| NM_004660.4 | NP_004651.2 | |||
| NR_136716.1 | ||||
| NR_136717.1 | 5 | 630 | ||
| NR_136718.1 | 5 | 630 | ||
| NR_136719.1 | ||||
| NR_136720.1 | ||||
| NR_136721.1 | ||||
| NR_136722.1 | 5 | 545 | ||
| NR_136723.1 | 5 | 545 | ||
| NR_136724.1 | ||||
| AF000984.1 | AAC51831.1 | |||
| AF000985.1 | AAC51832.1 | |||
| AK296366.1 | ||||
| AK302172.1 | ||||
| AK303176.1 | ||||
| AK303638.1 | ||||
| AK307884.1 | ||||
| AK310382.1 | ||||
| BC034942.2 | AAH34942.1 | |||
| DC394594.1 |
More Information
Set Membership: |
|

Genomic Map