Assay Details
Target Gene Details
Entrez Gene ID: | 84966 |
Gene Name: | immunoglobin superfamily member 21 |
Gene Aliases: |
- |
Location: |
Chr.1:18107746-18378483 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| IGSF21 | NM_032880.4 | NP_116269.3 | ||
| XM_011542319.2 | XP_011540621.1 | |||
| AK075316.1 | BAC11542.1 | |||
| AK294121.1 | ||||
| BC007618.1 | AAH07618.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv834313 | Chr.1:17955421 - 18162607 on Build GRCh38 | Loss |
|
| dgv177n54 | Chr.1:18093650 - 18117498 on Build GRCh38 | Loss |
|
| nsv1160182 | Chr.1:17655418 - 18282385 on Build GRCh38 | Duplication |
|
More Information
Additional Information:
For this assay, SNP(s) [rs544733989] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
|

Genomic Map