Assay Details
Target Gene Details
Entrez Gene ID: | 8514 |
Gene Name: | potassium voltage-gated channel subfamily A regulatory beta subunit 2 |
Gene Aliases: |
AKR6A5, HKvbeta2, HKvbeta2.1, HKvbeta2.2, KCNA2B, KV-BETA-2 |
Location: |
Chr.1:5992298-6101193 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| KCNAB2 | NM_001199861.1 | NP_001186790.1 | ||
| XM_005263514.2 | XP_005263571.1 | |||
| XM_011542321.2 | XP_011540623.1 | |||
| XM_017002619.1 | XP_016858108.1 | |||
| AK289819.1 | ||||
| AK290044.1 | ||||
| AK312395.1 | ||||
| AK315811.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv545173 | Chr.1:5833391 - 6025293 on Build GRCh38 | Gain |
|
| nsv1008071 | Chr.1:5892183 - 6084478 on Build GRCh38 | Loss |
|
| nsv953994 | Chr.1:5992941 - 6061340 on Build GRCh38 | Deletion |
|
| nsv1008999 | Chr.1:5996108 - 6094669 on Build GRCh38 | Gain |
|
| nsv831314 | Chr.1:5889409 - 6062997 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map