Assay Details
Target Gene Details
Entrez Gene ID: | 126755 |
Gene Name: | leucine rich repeat containing 38 |
Gene Aliases: |
- |
Location: |
Chr.1:13474977-13513747 on Build GRCh38 |
Assay Gene Location: | Within Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| LRRC38 | NM_001010847.1 | 2 | 1077 | NP_001010847.1 |
| BC016048.1 | 2 | 992 | AAH16048.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv3585258 | Chr.1:13459452 - 13496736 on Build GRCh38 | Loss |
|
| esv3891970 | Chr.1:13438356 - 13504122 on Build GRCh38 | Loss |
|
| esv3892081 | Chr.1:13472989 - 13621730 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map