Assay Details
Target Gene Details
Entrez Gene ID: | 100507670 |
Gene Name: | uncharacterized LOC100507670 |
Gene Aliases: |
- |
Location: |
Chr.1:151280024-151281929 on Build GRCh38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| LOC100507670 | NR_135595.1 | 1 | 134 | |
| BE385446.1 | 1 | 162 | ||
| DA674483.1 | 1 | 102 |
Target Gene Details
Entrez Gene ID: | 57592 |
Gene Name: | zinc finger protein 687 |
Gene Aliases: |
PDB6 |
Location: |
Chr.1:151281555-151291905 on Build GRCh38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ZNF687 | NM_001304764.1 | NP_001291693.1 | ||
| XM_005245366.3 | 1 | 239 | XP_005245423.1 | |
| CR749307.1 | CAH18162.1 | |||
| DQ655935.2 | 1 | 300 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv522840 | Chr.1:150961247 - 151375270 on Build GRCh38 | Gain |
|
| esv34461 | Chr.1:151268066 - 151439159 on Build GRCh38 | Gain |
|
| dgv2n68 | Chr.1:151136983 - 151309302 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map