Assay Details
Target Gene Details
Entrez Gene ID: | 65260 |
Gene Name: | cytochrome c oxidase assembly factor 7 (putative) |
Gene Aliases: |
C1orf163, RESA1, SELRC1 |
Location: |
Chr.1:52686342-52698366 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| COA7 | NM_023077.2 | NP_075565.2 | ||
| AK022501.1 | BAB14063.1 | |||
| AK225041.1 | ||||
| AK225087.1 | ||||
| BC015313.1 | AAH15313.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1001535 | Chr.1:52620368 - 53346539 on Build GRCh38 | Gain |
|
| esv2758934 | Chr.1:52660343 - 52823509 on Build GRCh38 | Loss |
|
More Information
Additional Information:
For this assay, SNP(s) [rs79150660] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
|

Genomic Map