Assay Details
Target Gene Details
Entrez Gene ID: | 699 |
Gene Name: | BUB1 mitotic checkpoint serine/threonine kinase |
Gene Aliases: |
BUB1A, BUB1L, hBUB1 |
Location: |
Chr.2:110637698-110678107 on Build GRCh38 |
Assay Gene Location: | Within Intron 16 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| BUB1 | NM_001278616.1 | NP_001265545.1 | ||
| NM_001278617.1 | NP_001265546.1 | |||
| NM_004336.4 | NP_004327.1 | |||
| AF011387.1 | AAC39546.1 | |||
| AF043294.2 | AAB97855.2 | |||
| AF046078.1 | AAC12729.1 | |||
| AF047471.1 | AAC03122.1 | |||
| AF053305.1 | AAC06259.1 | |||
| AK225317.1 | ||||
| AK291221.1 | ||||
| AK302418.1 | ||||
| BC017038.1 | ||||
| BC028201.1 | AAH28201.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv582696 | Chr.2:110610150 - 110711561 on Build GRCh38 | Gain |
|
| dgv730e214 | Chr.2:110395308 - 110848620 on Build GRCh38 | Gain |
|
| dgv7007n54 | Chr.2:110628656 - 110694962 on Build GRCh38 | Loss |
|
| nsv521956 | Chr.2:110634682 - 112341235 on Build GRCh38 | Loss |
|
| dgv4047n100 | Chr.2:110308696 - 112358403 on Build GRCh38 | Loss |
|
| esv3591965 | Chr.2:110642083 - 110829253 on Build GRCh38 | Gain |
|
More Information
Additional Information:
For this assay, SNP(s) [rs115805074] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
|

Genomic Map