Assay Details
Target Gene Details
Entrez Gene ID: | 3921 |
Gene Name: | ribosomal protein SA |
Gene Aliases: |
37LRP, 67LR, ICAS, LAMBR, LAMR1, LBP, LBP/p40, LRP, LRP/LR, NEM/1CHD4, SA, lamR, p40 |
Location: |
Chr.3:39406689-39412542 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| RPSA | NM_001304288.1 | NP_001291217.1 | ||
| NM_002295.5 | NP_002286.2 | |||
| BC066941.1 | AAH66941.1 | |||
| BC070263.1 | AAH70263.1 | |||
| X15005.1 | CAA33112.1 |
More Information
Additional Information:
For this assay, SNP(s) [rs61547423] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
|

Genomic Map