Assay Details
Target Gene Details
Entrez Gene ID: | 729085 |
Gene Name: | family with sequence similarity 198 member A |
Gene Aliases: |
C3orf41 |
Location: |
Chr.3:42979267-43057715 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| FAM198A | NM_001129908.2 | NP_001123380.2 | ||
| AK091001.1 | ||||
| AK311059.1 | ||||
| AK311069.1 | ||||
| DA262563.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv460523 | Chr.3:42984494 - 43021914 on Build GRCh38 | Loss |
|
| nsv590149 | Chr.3:42926105 - 43133715 on Build GRCh38 | Gain |
|
| esv2725206 | Chr.3:42997436 - 43005481 on Build GRCh38 | Deletion |
|

Genomic Map