Assay Details
Target Gene Details
Entrez Gene ID: | 3516 |
Gene Name: | recombination signal binding protein for immunoglobulin kappa J region |
Gene Aliases: |
AOS3, CBF1, IGKJRB, IGKJRB1, KBF2, RBP-J, RBPJK, RBPSUH, SUH, csl |
Location: |
Chr.4:26163489-26435131 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| RBPJ | XM_005248161.3 | XP_005248218.1 | ||
| XM_017008170.1 | XP_016863659.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv593847 | Chr.4:26130739 - 26218171 on Build GRCh38 | Loss |
|
| esv34177 | Chr.4:26038951 - 26206888 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map